Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Quinuclidinol, 99%, Thermo Scientific Chemicals

Catalog No. p-190157
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1619-34-7
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00151326
InChI Key IVLICPVPXWEGCA-UHFFFAOYSA-N
Synonym 3-quinuclidinol, quinuclidin-3-ol, 3-hydroxyquinuclidine, 1-azabicyclo 2.2.2 octan-3-ol, quinuclidinol, 1-aza-bicyclo 2.2.2 octan-3-ol, 3-hydroxy-1-azabicyclo 2.2.2 octane, 3-quinuclidinol dl-form, +--3-quinuclidinol, quinuclidine-3-ol
PubChem CID 15381
ChEBI CHEBI:115239
IUPAC Name 1-azabicyclo[2.2.2]octan-3-ol
SMILES C1CN2CCC1C(C2)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 217.0°C to 224.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Fieser 06,501
Merck Index 15, 8199
Solubility Information Solubility in water: soluble
Formula Weight 127.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Quinuclidinol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
EINECSNumber 216-578-4
RTECSNumber VD6191700
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.