Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193968 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 220210-56-0
Molecular Formula C11H15BO4
MDL Number MFCD01630855
InChI Key HVJDVHCPCSZDSR-UHFFFAOYSA-N
PubChem CID 2773299

Specifications

Melting Point 102°C
Color White
Quantity 1 g
Synonym 3-tert-butoxycarbonylphenylboronic acid, 3-tert-butoxycarbonyl phenylboronic acid, 3-t-butoxycarbonyl phenylboronic acid, tert-butyl 3-boronobenzoate, 3-tert-butoxycarbonyl phenyl boronic acid, 3-tert-butoxycarbonylphenyl boronic acid, 3-tert-butoxycarbonyl benzeneboronic acid, 3-t-butoxycarbonylphenylboronic acid, 3-2-methylpropan-2-yl oxycarbonyl phenyl boronic acid
SMILES CC(C)(C)OC(=O)C1=CC=CC(=C1)B(O)O
IUPAC Name {3-[(tert-butoxy)carbonyl]phenyl}boronic acid
Molecular Weight (g/mol) 222.05
Formula Weight 222.05
Physical Form Crystalline Powder
Chemical Name or Material 3-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.