Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%, Thermo Scientific Chemicals

Catalog No. p-4602069
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 642066-70-4
Molecular Formula C14H25BO4
Molecular Weight (g/mol) 268.16
MDL Number MFCD03788736
InChI Key MHSOBXCZCRNELG-UHFFFAOYNA-N
Synonym 4,4,5,5-tetramethyl-2-3-tetrahydro-2h-pyran-2-yl oxy prop-1-en-1-yl-1,3,2-dioxaborolane, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl oxy oxane, 4,4,5,5-tetramethyl-2-3-oxan-2-yloxy prop-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 53406456
IUPAC Name 4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-enyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=CCOC1CCCCO1

Specifications

Boiling Point 79°C (0.1 Torr)
Assay Percent Range 88% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 268.16
Percent Purity 90%
Chemical Name or Material 3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.