Learn More
Across International 30mm Diameter ID Dry Pressing Die Set
Supplier: Across International SDS30
A set of 30mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 30mm diameter cylinder-shape pellets.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.