Learn More
Uline Chrome Wire Shelving Unit - 30 x 18 x 54"
Supplier:  Uline H293954
Encompass
Wire Shelving Unit - 30 x 18 x 54", chrome-plated carbon steel, 800lbs shelf capacity (based on evenly distributed loads), overall capacity not to exceed 4,800lbs
Catalog No. NC2394741
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.