Learn More
Ace Glass, Inc. 355MM 14/20 3PRT MANIFOLD
355mm long double tube vacuum manifold with four #15 o-ring joint ends. Manifold has three stopcock ports with 14/20 inner joints and 0-10MM Easy Action stopcocks. Distance between ports is 100MM which will allow for flasks up to 250mL.