Learn More
MSC Fowler 54-422-450-1 0.05 dg Resolution Digital Combination Protractor and Inclinometer 0.2 dg Accuracy, 360 dg Range, Magnetic Base
Fowler 54-422-450-1 0.05 dg Resolution Digital Combination Protractor and Inclinometer 0.2 dg Accuracy, 360 dg Range, Magnetic Base