Learn More
3B Scientific™ Dismantlable Transformer
SDPSCI_ED
For the generation of a powerful magnetic flux.
$31.25
Specifications
For Use With (Application) Generation of a powerful magnetic flux
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S97702
SDPSCI_ED
View Documents
3B Scientific™
U8497470
Metal Ring
Each of 1 for $31.25
Add to Cart
Description
Description
  • Made of shock-proof plastic
  • Technical data printed on coils: number of turns, maximum continuous current, effective resistance and inductance
  • Connections via 4-mm safety sockets
  • Dimensions: 4.8 × 3.6 × 2.8 in. (120 × 90 × 70mm)
  • Opening for transformer cores: 1.68 × 1.68 in. (42 × 42mm)
Specifications
Specifications
Generation of a powerful magnetic flux
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.