Learn More
3B Scientific™ Life/form™ Episiotomy Suturing Simulators
SDPSCI_ED
Portable, lifelike textured simulator provides realistic way for students to learn good surgical techniques.
$465.00 - $504.00
Specifications
Includes Instruction booklet
View More Specs
Includes: Instruction booklet
Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S07990
SDPSCI_ED
View Documents
3B Scientific™
W44096RM
Right Mediolateral Episiotomy Suturing Simulator
Each for $504.00
missing translation for 'QuantityLeft'
Add to Cart
S07988
SDPSCI_ED
View Documents
3B Scientific™
W44415
Midline Episiotomy Suturing Simulator
Each for $465.00
missing translation for 'QuantityLeft'
Add to Cart
S07989
SDPSCI_ED
View Documents
3B Scientific™
W44416
Left Mediolateral Episiotomy Suturing Simulator
Each for $465.00
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Provides students with variety of repair experiences without constraint of time and concern for safety
  • Students can use in learning lab with an instructor, or individually in clinical setting just prior to patient experience
  • Use as homework teaching aid that can be signed out at night and returned next day
  • Learner develop “feel” for instrument handling, tension on suture and advantages of one method of tying knots over another
Warranty and Services
Three years
Specifications
Specifications
Instruction booklet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.