Learn More
Ace Glass, Inc. 3000mL jacketed filter reactor, complete with #15 Ace-Thred inlet/outlets, 150mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings
Supplier:  Ace Glass, Inc. 6386460
Encompass_Preferred
3000mL jacketed filter reactor, complete with #15 Ace-Thred inlet/outlets, 150mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings
Catalog No. 50-972-814
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.