Learn More
RS Hughes Inc 3M EMI 1181 1 X 18YD COPPER FL
SDP
Supplier:  RS Hughes Inc 05400727468
Encompass_Preferred
3M Copper Tape - 1 in Width x 2.6 mil Total Thickness - 27468
Catalog No. NC9751620
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.