Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ H-Series Respirator Hood

Replacement respirator hood for 3M™ Hood Assemblies H-421 and H-422.

Supplier:  3M™ 7000002073

Catalog No. 19-088-1289

This item is not returnable. View return policy

Description

Description

  • Lightweight, comfortable
  • Provides head, neck and respiratory protection from pesticide and chemical overspray
  • Loose fitting
  • For use with belt-mounted powered air purifying respirators or supplied air components
  • Color: White

3M H-400 Series Hoods

  • Feature lightweight Tychem™ QC Fabric and provide high levels of airflow for maximum comfort For pharmaceutical, laboratory, and health care applications as well as during painting and sanding operations H-400 is available with hardhat and choice of inner shroud or collar
  • 3M Catalog# H-420-10

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Specifications

Specifications

1,000
White
Respirator Hood
Belt mounted powered air purifying respirator and supplied air respirator system
H-420-10
NIOSH
Tychem QC
Machine operation, military maintenance, repair and operation, pharmaceutical
Two hoods, cap or hardhat suspension, chin strap, two faceshield covers and a plastic squeeze clamp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.