Learn More
Sklar Instruments Single Use, Econo-Sterile, Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, Sterile, CS of 100. .
Supplier: Sklar Instruments 962650
Single Use, Econo-Sterile, Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, Sterile, CS of 100. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.