Learn More
Sklar Instruments Scissors, Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Angled, Smooth, Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 642142
Encompass_Preferred
Scissors, Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Angled, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms, and in procedures involving fine tissue such as OB/GYN and dermatology. This product is angled with smooth, sharp/sharp tips and a length of 4-1/2 inches.
Catalog No. 50-109-3950
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.