Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Pyrenyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193882 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 872050-52-7
Molecular Formula C22H15BO2
Molecular Weight (g/mol) 322.17
MDL Number MFCD28130386
InChI Key ISMRPUXCQLIMJL-UHFFFAOYSA-N
Synonym 4-(1-Pyrenyl)benzeneboronic Acid
PubChem CID 59217173
IUPAC Name [4-(pyren-1-yl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Quantity 1 g
Formula Weight 322.17
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Pyrenyl)phenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.