Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97+%, Thermo Scientific™

Catalog No. p-4510681
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 859850-94-5
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
InChI Key XEPIORKIFKJATD-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazolylmethyl benzaldehyde, 4-1h-1,2,4-triazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-1,2,4-triazol-1-ylmethyl
PubChem CID 7164655
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC=C1CN2C=NC=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.