Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Bromoethyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7119372 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 52062-92-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016541
InChI Key BKMRWJWLBHHGCF-UHFFFAOYSA-N
PubChem CID 104068
IUPAC Name 4-(2-bromoethyl)benzoic acid
SMILES OC(=O)C1=CC=C(CCBr)C=C1

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Bromoethyl)benzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.