Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline hydrochloride, 97%, Thermo Scientific™

Catalog No. MO08102DE
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

113674-95-6

Molecular Formula

C13H9Cl2F4NO

Molecular Weight (g/mol)

342.115

MDL Number

MFCD09702423

InChI Key

BBBUNESZGCZWJR-UHFFFAOYSA-N

Synonym

4-2-chloro-4-trifluoromethyl phenoxy-2-fluoroaniline hydrochloride, 4-2-chloro-4-trifluoromethyl phenoxy-2-fluoroaniline-hydrogen chloride 1/1

PubChem CID

18525688

IUPAC Name

4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline;hydrochloride

SMILES

C1=CC(=C(C=C1C(F)(F)F)Cl)OC2=CC(=C(C=C2)N)F.Cl

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

5 g

Formula Weight

342.12

Percent Purity

97%

Chemical Name or Material

4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline hydrochloride

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes damage to organs through prolo

Precautionary Statement

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.