Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(2-Hydroxyethyl)benzonitrile, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CNC6H4(CH2)2OH |
Quantity | 1 g |
Formula Weight | 147.18 |
Percent Purity | 97% |
Chemical Name or Material | 4-(2-Hydroxyethyl)benzonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction