Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Methyl-1H-imidazol-1-yl)benzylamine, 90%, Thermo Scientific™

Catalog No. p-3527252
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 883291-45-0
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD06617944
InChI Key JOMOQABDRBJMDQ-UHFFFAOYSA-N
Synonym 4-2-methyl-1h-imidazol-1-yl benzylamine, 1-4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazol-1-yl phenyl methanamine, benzenemethanamine,4-2-methyl-1h-imidazol-1-yl, 4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazolyl phenyl methylamine, 1-4-2-methylimidazol-1-yl phenyl methanamine
PubChem CID 4913134
IUPAC Name [4-(2-methylimidazol-1-yl)phenyl]methanamine
SMILES CC1=NC=CN1C2=CC=C(C=C2)CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 90%
Chemical Name or Material 4-(2-Methyl-1H-imidazol-1-yl)benzylamine
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.