Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4′-Bis(decyloxy)-3-methylazobenzene 95.0+%, TCI America™
SDP

Catalog No. p-7123635 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
500 mg
5 g

Chemical Identifiers

CAS 1627844-78-3
Molecular Formula C33H52N2O2
Molecular Weight (g/mol) 508.79
MDL Number MFCD30742892
InChI Key HBJQHHIKMMWGLL-UHFFFAOYSA-N
PubChem CID 132274868
IUPAC Name [4-(decyloxy)-3-methylphenyl][4-(decyloxy)phenyl]diazene
SMILES CCCCCCCCCCOC1=CC=C(C=C1)N=NC1=CC(C)=C(OCCCCCCCCCC)C=C1

Specifications

Quantity 500 mg
Formula Weight 508.79
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(decyloxy)-3-methylazobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.