Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4′-Carbonyldimorpholine 98.0+%, TCI America™
SDP

Catalog No. p-7125747 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 38952-62-4
Molecular Formula C9H16N2O3
Molecular Weight (g/mol) 200.24
MDL Number MFCD00593814
InChI Key QONGECDDDTYBGS-UHFFFAOYSA-N
PubChem CID 655428
IUPAC Name 4-(morpholine-4-carbonyl)morpholine
SMILES O=C(N1CCOCC1)N1CCOCC1

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Carbonyldimorpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.