Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. p-7125275 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 70291-67-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.662
InChI Key KDWPYZPZUPWJGB-UHFFFAOYSA-N
PubChem CID 10420251
IUPAC Name 4-(4-chlorophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)Cl

Specifications

Melting Point 73°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 197.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.