Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4′-Dichlorochalcone 98.0+%, TCI America™
SDP

Catalog No. p-7128500 missing translation for 'brandsLinkText'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g

Chemical Identifiers

CAS 19672-59-4
Molecular Formula C15H10Cl2O
Molecular Weight (g/mol) 277.14
MDL Number MFCD00018704
InChI Key YMEMCRBNZSLQCQ-XCVCLJGOSA-N
Synonym 4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
PubChem CID 5377011
IUPAC Name (2E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
SMILES ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 158°C
Color Yellow
Quantity 5 g
Formula Weight 277.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorochalcone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.