Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, 97%, Thermo Scientific™

Catalog No. p-4509924
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 439691-80-2
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
InChI Key DJJFKXUSAXIMLS-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzaldehyde, 1-4-formylbenzyl-4-methylpiperazine, benzaldehyde,4-4-methyl-1-piperazinyl methyl, 4-4-methylpiperazino methyl benzaldehyde, 4-4-methylpiperazinyl methyl benzaldehyde, 4-4-methyl piperazin-1-ylmethyl benzaldehyde, 4-4-methyl-1-piperazinyl methyl benzaldehyde
PubChem CID 7164651
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 218.3
Percent Purity 97%
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.