Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(4-Morpholinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC431770500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 480424-75-7, 109-99-9
Molecular Formula C11H14BrMgNO
Molecular Weight (g/mol) 280.45
MDL Number MFCD03840890
InChI Key OFPHGAKUXVPRKI-UHFFFAOYSA-M
Synonym bromo 4-4-morpholinylmethyl phenyl magnesium, 4-4-bromomagnesio phenyl methyl morpholine, ofphgakuxvprki-uhfffaoysa-m, 4-4-morpholinylmethyl phenyl magnesiu, 4-4-morpholinylmethyl phenyl magnesium bromide, 4-4-morpholino methyl phenylmagnesium bromide, 4-4-morpholinylmethyl phenyl magnesium bromide solution, 4-4-morpholinylmethyl phenyl magnesium bromide solution, 0.25 m in thf, 4-4-morpholino methyl phenylmagnesium bromide 0.25 m in tetrahydrofuran, 4-4-morpholino methyl phenylmagnesium bromide, 0.25m in tetrahydrofuran, grignard reagent
PubChem CID 24722692
IUPAC Name magnesium;4-(phenylmethyl)morpholine;bromide
SMILES Br[Mg]C1=CC=C(CN2CCOCC2)C=C1

Specifications

Color Yellow
CAS Min % 4.54, 89.4
CAS Max % 10.6, 95.46
Physical Form Solution
Quantity 50 mL
Flash Point −17°C
Solubility Information Solubility in water: reacts
Formula Weight 280.44
Concentration or Composition (by Analyte or Components) 0.15 to 0.35M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.926
Density 0.9260g/mL
Chemical Name or Material [4-(4-Morpholinylmethyl)phenyl]magnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.