Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dihydro-2-phenyloxazole 98.0+%, TCI America™
SDP

Catalog No. p-7127140 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7127-19-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00075541
InChI Key ZXTHWIZHGLNEPG-UHFFFAOYSA-N
Synonym (4,5-Dihydro-2-oxazolyl)benzene, 2-Phenyl(2-oxazoline)
PubChem CID 244030
IUPAC Name 2-phenyl-4,5-dihydro-1,3-oxazole
SMILES C1CN=C(O1)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 243°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4,5-Dihydro-2-phenyloxazole
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.