Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine 98.0+%, TCI America™
SDP

Catalog No. p-7118048 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 5334-30-5
Molecular Formula C11H9N5
Molecular Weight (g/mol) 211.23
MDL Number MFCD00022901
InChI Key KKDPIZPUTYIBFX-UHFFFAOYSA-N
Synonym 4-amino-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-amine, pp3, pp 3, 1-phenylpyrazolo 3,4-d pyrimidin-4-amine, chembl65063, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 4-amino-7-phenylpyrazol 3,4-d pyrimidine, 4-amino-1-phenylpyrazolo 3,4-e pyrimidine, 1h-pyrazolo 3,4-d pyrimidin-4-amine, 1-phenyl
PubChem CID 4879
IUPAC Name 1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES NC1=C2C=NN(C2=NC=N1)C1=CC=CC=C1

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 200 mg
Formula Weight 211.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.