Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-methoxybenzonitrile 97.0+%, TCI America™
SDP

Catalog No. p-7118072 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 7251-09-4
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
MDL Number MFCD09027303
InChI Key KTDRJLRJAHBQDQ-UHFFFAOYSA-N
Synonym 4-Cyano-3-methoxyaniline, 5-Amino-2-cyanoanisole
PubChem CID 232735
IUPAC Name 4-amino-2-methoxybenzonitrile
SMILES COC1=C(C=CC(=C1)N)C#N

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 148.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methoxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.