Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-chlorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. A19715G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 2486-71-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00460423
InChI Key YIYBPEDZAUFQLO-UHFFFAOYSA-N
Synonym 4-amino-3-chloro-benzoic acid, 3-chloro-4-aminobenzoic acid, 4-amino-3-chlorbenzoic acid, benzoic acid, 4-amino-3-chloro, 4-amino-3-chlorobenzoicacid, 4-amino-3-chloro-benzoicacid, 4-amnio-3-chlorobenzioc acid, pubchem4652, acmc-209gf7, ksc494k4j
PubChem CID 17211
IUPAC Name 4-amino-3-chlorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)Cl)N

Specifications

Melting Point 228°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-chlorobenzoic Acid
EINECSNumber (3)-1480
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.