Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-(trifluoromethoxy)benzoic acid, 95%, Thermo Scientific™

Catalog No. TL00679DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 175278-22-5
Molecular Formula C8H6F3NO3
Molecular Weight (g/mol) 221.135
InChI Key IXJFWBNYFTWBOR-UHFFFAOYSA-N
Synonym 4-amino-3-trifluoromethoxy benzoic acid, rarechem al bo 0798, buttpark 83\07-32, 4-amino-3-trifluoromethoxy-benzoic acid, 4-amine-3-trifluoromethoxy benzoic acid, benzoic acid, 4-amino-3-trifluoromethoxy, 4-amino-3-trifluoromethoxybenzoicacid, pubchem14042, acmc-209e9s
PubChem CID 2735951
IUPAC Name 4-amino-3-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)OC(F)(F)F)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.13
Percent Purity 95%
Chemical Name or Material 4-Amino-3-(trifluoromethoxy)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.