Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-(trifluoromethoxy)benzonitrile, ≥97%, Thermo Scientific™

Catalog No. TL00680EA
Click to view available options
Quantity:

Chemical Identifiers

CAS

175278-23-6

Molecular Formula

C8H5F3N2O

Molecular Weight (g/mol)

202.14

MDL Number

MFCD00204177

InChI Key

QNJPSMLILRHZOW-UHFFFAOYSA-N

Synonym

4-amino-3-trifluoromethoxy benzonitrile, benzonitrile, 4-amino-3-trifluoromethoxy, 3-trifluoromethoxy-4-aminobenzonitrile, 4-amino-3-trifluoromethoxy benzenecarbonitrile, pubchem4688, ksc535q4r, buttpark 81\01-97, 4-cyano-2-trifluoromethoxy-aniline, 4-cyano-2-trifluoromethoxy aniline, 4-amino-3-trifluoromethoxy-benzonitrile

PubChem CID

2735952

IUPAC Name

4-amino-3-(trifluoromethoxy)benzonitrile

SMILES

NC1=C(OC(F)(F)F)C=C(C=C1)C#N

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Packaging

Amber glass bottle

Quantity

10 g

Formula Weight

202.13

Percent Purity

97%

Chemical Name or Material

4-Amino-3-(trifluoromethoxy)benzonitrile

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

Precautionary Statement

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.