Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7118144 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 123654-26-2
Molecular Formula C9H8ClNO3
Molecular Weight (g/mol) 213.617
MDL Number MFCD12923204
InChI Key KRMUVKSAOVLXLF-UHFFFAOYSA-N
Synonym 4-Amino-5-chlorocoumaran-7-carboxylic Acid
PubChem CID 10632401
IUPAC Name 4-amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES C1COC2=C1C(=C(C=C2C(=O)O)Cl)N

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 200 mg
Formula Weight 213.62
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.