Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyl-2-hydroxymorpholin-3-one 98.0+%, TCI America™
SDP

Catalog No. p-7123838 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 287930-73-8
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD07776435
InChI Key CAGSEMPCDRYWFN-UHFFFAOYSA-N
PubChem CID 9942497
IUPAC Name 4-benzyl-2-hydroxymorpholin-3-one
SMILES C1COC(C(=O)N1CC2=CC=CC=C2)O

Specifications

Melting Point 136°C
Color White
Quantity 1 g
Formula Weight 207.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl-2-hydroxymorpholin-3-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.