Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. p-7119015 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 52805-36-4
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00079701
InChI Key UDAOJHAASAWVIQ-UHFFFAOYSA-N
Synonym 4-benzyloxybenzonitrile, 4-benzyloxy benzonitrile, 4-benzyloxy-benzonitrile, benzonitrile, 4-phenylmethoxy, 4-phenylmethoxy benzenecarbonitrile, maybridge1_005982, benzonitrile analogue, 5a, acmc-209l1v, 4-cyanophenoxy phenyl methane
PubChem CID 561371
IUPAC Name 4-phenylmethoxybenzonitrile
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)C#N

Specifications

Melting Point 94°C
Color White
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.