Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,5-difluoroaniline 98.0+%, TCI America™
SDP

Catalog No. p-7119462 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 112279-60-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.006
MDL Number MFCD01861125
InChI Key XOYHFIQPPOJMFK-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoro-phenylamine, 4-bromo-2,5-difluoro-aniline, 4-bromo-2,5-difluorophenylamine, pubchem2928, acmc-1bvn4, ksc494m7l, 4-bromo-2,5-difluorobenzenamine, 2,5-difluoro-4-bromoaniline, 4-bromo-2,5-difluoroaniline, benzenamine, 4-bromo-2,5-difluoro
PubChem CID 2773285
IUPAC Name 4-bromo-2,5-difluoroaniline
SMILES C1=C(C(=CC(=C1F)Br)F)N

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-difluoroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.