Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,5-difluorobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B23881G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 207974-14-9
Molecular Formula C6H2BrClF2O2S
Molecular Weight (g/mol) 291.49
MDL Number MFCD00042181
InChI Key SBMKFWMFNIEPDN-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-2,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, 4-bromo-3,6-difluoro-benzenesulfonylchloride, pubchem5058, acmc-209feg, 4-bromo-2,5-difluorobenzenesulfonylchloride
PubChem CID 574590
IUPAC Name 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride
SMILES FC1=CC(=C(F)C=C1Br)S(Cl)(=O)=O

Specifications

Melting Point 41°C
Quantity 1 g
UN Number 3261
Formula Weight 291.49
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-difluorobenzenesulfonyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.