Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Bromo-2,6-dimethylaniline 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Melting Point | 51°C |
Quantity | 25 g |
Physical Form | Crystal-Powder at 20°C |
Formula Weight | 200.08 |
Percent Purity | ≥98.0% (GC,T) |
Chemical Name or Material | 4-Bromo-2,6-dimethylaniline |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction