Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. p-7119363 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 76283-09-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00055467
InChI Key XMHNLZXYPAULDF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl bromide, 4-bromo-1-bromomethyl-2-fluorobenzene, 2-fluoro-4-bromobenzyl bromide, 4-bromo-2-fluorobenzylbromide, 2-fluoro-4-bromo-benzylbromide, alpha,4-dibromo-2-fluorotoluene, unii-0z26xyc1jy, 4,alpha-dibromo-2-fluorotoluene, 4-bromo-1-bromomethyl-2-fluoro-benzene, 0z26xyc1jy
PubChem CID 2733660
IUPAC Name 4-bromo-1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)CBr

Specifications

Melting Point 37°C
Quantity 5 g
UN Number 2923
Formula Weight 267.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzyl Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.