Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. p-7123742 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 128071-98-7
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD04112504
InChI Key PTPTZLXZHPPVKG-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromopyridine, 4-bromo-2-fluoro-pyridine, pyridine, 4-bromo-2-fluoro, abbypharma ap-17-5099, pubchem1228, acmc-209bdf, pyff21, 4-bromo-2-fluoro pyridine, ksc493i4r, 4-bromanyl-2-fluoranyl-pyridine
PubChem CID 2762851
IUPAC Name 4-bromo-2-fluoropyridine
SMILES C1=CN=C(C=C1Br)F

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluoropyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.