Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-iodophenol 98.0+%, TCI America™
SDP

Catalog No. p-7123864 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 207115-22-8
Molecular Formula C6H4BrIO
Molecular Weight (g/mol) 298.905
MDL Number MFCD09029644
InChI Key UXIULWIJWDJDQD-UHFFFAOYSA-N
PubChem CID 10924532
IUPAC Name 4-bromo-2-iodophenol
SMILES C1=CC(=C(C=C1Br)I)O

Specifications

Melting Point 75°C
Quantity 1 g
Formula Weight 298.91
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-iodophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.