Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-bromo-3-methyl-1H-pyrazol-5-amine hydrobromide, Thermo Scientific™

Catalog No. KM01090DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 167683-86-5
Molecular Formula C4H7Br2N3
Molecular Weight (g/mol) 256.929
InChI Key UGCOULAJNKWOPX-UHFFFAOYSA-N
Synonym 5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide
PubChem CID 2778427
IUPAC Name 4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide
SMILES CC1=C(C(=NN1)N)Br.Br

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 256.93
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-methyl-1H-pyrazol-5-amine hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.