Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromobutyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123897 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 1190-14-3
Molecular Formula C4H10BrO3P
Molecular Weight (g/mol) 217.00
MDL Number MFCD00466589
InChI Key FXFARGAYPHUQQL-UHFFFAOYSA-N
PubChem CID 18409311
IUPAC Name (4-bromobutyl)phosphonic acid
SMILES OP(O)(=O)CCCCBr

Specifications

Melting Point 129°C
Color White
Quantity 200 mg
UN Number 3261
Formula Weight 217.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromobutyl)phosphonic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.