Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(Bromomethyl)-2,1,3-benzoxadiazole, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 213.03 |
Percent Purity | 95% |
Chemical Name or Material | 4-(Bromomethyl)-2, 1, 3-benzoxadiazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction