Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromophenyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7123898 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 16839-13-7
Molecular Formula C6H6BrO3P
Molecular Weight (g/mol) 236.989
MDL Number MFCD00159409
InChI Key XDGIQCFWQNHSMV-UHFFFAOYSA-N
PubChem CID 95011
IUPAC Name (4-bromophenyl)phosphonic acid
SMILES C1=CC(=CC=C1P(=O)(O)O)Br

Specifications

Melting Point 202°C
Quantity 200 mg
Formula Weight 236.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromophenyl)phosphonic Acid
EINECSNumber (3)-2524
RTECSNumber SZ6870300
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.