Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenylacetone 98.0+%, TCI America™
SDP

Catalog No. p-7123508 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 6186-22-7
Molecular Formula C24H24N2O5S
Molecular Weight (g/mol) 452.53
MDL Number MFCD00210401
InChI Key CDUPPXGKUKTGLU-UHFFFAOYSA-N
Synonym 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
PubChem CID 736332
IUPAC Name methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate
SMILES COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromophenylacetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.