Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. p-7123224 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 37909-95-8
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00100665
InChI Key JZDZRFNMDCBTNS-UHFFFAOYSA-N
Synonym 4-butyl-1,1'-biphenyl, 4-butylbiphenyl, 4-n-butylbiphenyl, 1,1'-biphenyl, 4-butyl, 4-butyl biphenyl, unii-1ty72t98fq, 1-p-biphenylylbutane, biphenyl, 4-butyl, 1-butyl-4-phenyl-benzene, 4-butyldiphenyl
PubChem CID 2775237
IUPAC Name 1-butyl-4-phenylbenzene
SMILES CCCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 129°C
Quantity 5 g
Formula Weight 210.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Butylbiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.