Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2,1,3-benzoxadiazole 98.0+%, TCI America™
SDP

Catalog No. p-7124921 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7116-16-7
Molecular Formula C6H3ClN2O
Molecular Weight (g/mol) 154.553
MDL Number MFCD00191329
InChI Key YLEBWUGKYMZPQQ-UHFFFAOYSA-N
Synonym 4-Chlorobenzofurazan
PubChem CID 592984
IUPAC Name 4-chloro-2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C(=C1)Cl

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
Formula Weight 154.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2,1,3-benzoxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.