Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Chloro-3-fluoroanisole 95.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 196°C |
Quantity | 5 g |
Formula Weight | 160.57 |
Percent Purity | ≥95.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 4-Chloro-3-fluoroanisole |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction