Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, ∽98 to 99%, MP Biomedicals™

Catalog No. ICN10136201 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

10199-89-0

Molecular Formula

C6H2ClN3O3

Molecular Weight (g/mol)

199.55

InChI Key

IGHBXJSNZCFXNK-UHFFFAOYSA-N

Synonym

4-chloro-7-nitrobenzofurazan, nbd chloride, 4-chloro-7-nitrobenzo-2-oxa-1,3-diazole, nbd-cl, 7-chloro-4-nitrobenzofurazan, 2,1,3-benzoxadiazole, 4-chloro-7-nitro, nbd-chloride, nbd-c 1, 1-chloro-4-nitrobenzoxadiazole, 4-nitro-7-chlorobenzofurazan

PubChem CID

25043

ChEBI

CHEBI:78878

IUPAC Name

4-chloro-7-nitro-2,1,3-benzoxadiazole

SMILES

C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

Assay Percent Range

Approx. 98 to 99%

Quantity

1 g

Formula Weight

199.6

Chemical Name or Material

4-Chloro-7-Nitrobenz-2-Oxa-1, 3-Diazole

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.